missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-valine, 98+%

Catalog No. p-7044955
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0885603 1 g
AAL0885606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0885603 Supplier Thermo Scientific Chemicals Supplier No. L0885603
Only null left

CAS: 1685-33-2 | C13H17NO4 | 251.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1685-33-2
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00065703
InChI Key CANZBRDGRHNSGZ-LLVKDONJSA-N
Synonym z-d-val-oh, cbz-d-valine, n-benzyloxycarbonyl-d-valine, cbz-d-val-oh, n-cbz-d-valine, z-d-valine, n-carbobenzoxy-d-valine, benzyloxycarbonyl-d-valine, r-2-benzyloxy carbonyl amino-3-methylbutanoic acid, d-valine, n-phenylmethoxy carbonyl
PubChem CID 712434
IUPAC Name (2R)-3-methyl-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CC(C)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 58°C to 60°C
Quantity 1 g
Beilstein 2056616
Optical Rotation +3.8° (c=2 in Acetic Acid)
Formula Weight 251.28
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-valine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only