missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-methionine, 98%

Catalog No. AAH6315206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6315206 5 g
AAH6315214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6315206 Supplier Thermo Scientific Chemicals Supplier No. H6315206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 28862-80-8 | C13H17NO4S | 283.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28862-80-8
Molecular Formula C13H17NO4S
Molecular Weight (g/mol) 283.34
MDL Number MFCD00026043
InChI Key FPKHNNQXKZMOJJ-LLVKDONJSA-N
Synonym z-d-met-oh, n-cbz-d-methionine, cbz-d-methionine, z-d-methionine, n-carbobenzoxy-d-methionine, r-2-benzyloxy carbonyl amino-4-methylthio butanoic acid, z-l-methionine, carbobenzyloxy-d-methionine, 2r-2-benzyloxy carbonyl amino-4-methylsulfanyl butanoic acid, pubchem14967
PubChem CID 1712153
IUPAC Name (2R)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CSCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 68°C to 72°C (decomposition)
Beilstein 2058695
Quantity 5 g
Formula Weight 283.24
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-D-methionine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only