missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%
CAS: 1947-00-8 | C14H19NO4 | 265.309 g/mol
$100.99 - $278.92
Chemical Identifiers
| CAS | 1947-00-8 |
|---|---|
| Molecular Formula | C14H19NO4 |
| Molecular Weight (g/mol) | 265.309 |
| MDL Number | MFCD00004423 |
| InChI Key | RXQDBVWDABAAHL-UHFFFAOYSA-N |
| Synonym | z-6-aminohexanoic acid, z-epsilon-acp-oh, 6-benzyloxy carbonyl amino hexanoic acid, n-benzyloxycarbonyl-6-aminocaproic acid, cbz-, a-aminocaproic acid, n-cbz-6-aminohexanoic acid, n-benzyloxycarbonyl-6-aminohexanoic acid, n-carbobenzoxy-epsilon-aminocaproic acid, 6-phenylmethoxy carbonyl amino hexanoic acid |
| PubChem CID | 74758 |
| IUPAC Name | 6-(phenylmethoxycarbonylamino)hexanoic acid |
| SMILES | C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0899706
|
Thermo Scientific Chemicals
L0899706 |
5 g |
Each for $100.99
|
|
|||||
|
AAL0899714
|
Thermo Scientific Chemicals
L0899714 |
25 g |
Each for $278.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1947-00-8 | |
| 265.309 | |
| RXQDBVWDABAAHL-UHFFFAOYSA-N | |
| 74758 | |
| C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O |
| C14H19NO4 | |
| MFCD00004423 | |
| z-6-aminohexanoic acid, z-epsilon-acp-oh, 6-benzyloxy carbonyl amino hexanoic acid, n-benzyloxycarbonyl-6-aminocaproic acid, cbz-, a-aminocaproic acid, n-cbz-6-aminohexanoic acid, n-benzyloxycarbonyl-6-aminohexanoic acid, n-carbobenzoxy-epsilon-aminocaproic acid, 6-phenylmethoxy carbonyl amino hexanoic acid | |
| 6-(phenylmethoxycarbonylamino)hexanoic acid |
Specifications
| 1947-00-8 | |
| C14H19NO4 | |
| 5 g | |
| z-6-aminohexanoic acid, z-epsilon-acp-oh, 6-benzyloxy carbonyl amino hexanoic acid, n-benzyloxycarbonyl-6-aminocaproic acid, cbz-, a-aminocaproic acid, n-cbz-6-aminohexanoic acid, n-benzyloxycarbonyl-6-aminohexanoic acid, n-carbobenzoxy-epsilon-aminocaproic acid, 6-phenylmethoxy carbonyl amino hexanoic acid | |
| C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O | |
| 265.309 | |
| 265.31 | |
| N-Benzyloxycarbonyl-6-aminohexanoic acid |
| 53°C to 57°C | |
| MFCD00004423 | |
| 2288508 | |
| RXQDBVWDABAAHL-UHFFFAOYSA-N | |
| 6-(phenylmethoxycarbonylamino)hexanoic acid | |
| 74758 | |
| 99% |
Safety and Handling
EINECSNumber : 217-748-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only