missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-2-methylalanine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6311906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 65°C to 69°C (decomposition) | |
| 15030-72-5 | |
| C12H15NO4 | |
| 2218009 | |
| QKVCSJBBYNYZNM-UHFFFAOYSA-N | |
| 2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid | |
| 237.255 | |
| 237.25 | |
| Powder |
| N-Benzyloxycarbonyl-2-methylalanine | |
| White | |
| MFCD00004190 | |
| z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl | |
| CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 294936 | |
| 98% |
Chemical Identifiers
| 15030-72-5 | |
| 237.255 | |
| QKVCSJBBYNYZNM-UHFFFAOYSA-N | |
| 294936 | |
| CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
| C12H15NO4 | |
| MFCD00004190 | |
| z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl | |
| 2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only