missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-2-methylalanine, 98%, Thermo Scientific Chemicals

$147.11 - $147.11

Chemical Identifiers

CAS 15030-72-5
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00004190
InChI Key QKVCSJBBYNYZNM-UHFFFAOYSA-N
Synonym z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl
PubChem CID 294936
IUPAC Name 2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6311906
View Documents
Thermo Scientific Chemicals
H6311906
5 g
Each for $147.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15030-72-5
237.255
QKVCSJBBYNYZNM-UHFFFAOYSA-N
294936
CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1
C12H15NO4
MFCD00004190
z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl
2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

65°C to 69°C (decomposition)
White
MFCD00004190
z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl
CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1
5 g
294936
98%
N-Benzyloxycarbonyl-2-methylalanine
15030-72-5
C12H15NO4
2218009
QKVCSJBBYNYZNM-UHFFFAOYSA-N
2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid
237.255
237.25
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only