missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzylmaleimide 98.0+%, TCI America™
Supplier: TCI America B180525G
Specifications
| N-Benzylmaleimide | |
| 70°C | |
| 177°C | |
| MFCD00014540 | |
| 1759 | |
| MKRBAPNEJMFMHU-UHFFFAOYSA-N | |
| 1-benzyl-2,5-dihydro-1H-pyrrole-2,5-dione | |
| 74204 | |
| ≥98.0% (HPLC) |
| 1631-26-1 | |
| White | |
| C11H9NO2 | |
| 25 g | |
| n-benzylmaleimide, 1-benzyl-1h-pyrrole-2,5-dione, n-benzyl maleimide, 1h-pyrrole-2,5-dione, 1-phenylmethyl, 1-benzylmaleimide, n-benzylmaleinimide, 1-benzyl-pyrrole-2,5-dione, n-benzyl-maleimide, 1-benzyl-2,5-dihydro-1h-pyrrole-2,5-dione, n-benzylmaleimide dihydrate | |
| O=C1C=CC(=O)N1CC1=CC=CC=C1 | |
| 187.20 | |
| 187.20 | |
| Crystalline Powder |
Chemical Identifiers
| 1631-26-1 | |
| 187.20 | |
| MKRBAPNEJMFMHU-UHFFFAOYSA-N | |
| 74204 | |
| O=C1C=CC(=O)N1CC1=CC=CC=C1 |
| C11H9NO2 | |
| MFCD00014540 | |
| n-benzylmaleimide, 1-benzyl-1h-pyrrole-2,5-dione, n-benzyl maleimide, 1h-pyrrole-2,5-dione, 1-phenylmethyl, 1-benzylmaleimide, n-benzylmaleinimide, 1-benzyl-pyrrole-2,5-dione, n-benzyl-maleimide, 1-benzyl-2,5-dihydro-1h-pyrrole-2,5-dione, n-benzylmaleimide dihydrate | |
| 1-benzyl-2,5-dihydro-1H-pyrrole-2,5-dione |
Safety and Handling
EINECSNumber : (5)-6426
TSCA : No