accessibility menu, dialog, popup

UserName

OON

CAS RN 1631-26-1

IUPAC Name: 1-benzyl-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-benzylpyrrole-2,5-dione
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: MKRBAPNEJMFMHU-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results