missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylmaleimide 98.0+%, TCI America™

Catalog No. B18055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18055G 5 g
B180525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18055G Supplier TCI America Supplier No. B18055G

Chemical Identifiers

CAS 1631-26-1
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD00014540
InChI Key MKRBAPNEJMFMHU-UHFFFAOYSA-N
Synonym n-benzylmaleimide, 1-benzyl-1h-pyrrole-2,5-dione, n-benzyl maleimide, 1h-pyrrole-2,5-dione, 1-phenylmethyl, 1-benzylmaleimide, n-benzylmaleinimide, 1-benzyl-pyrrole-2,5-dione, n-benzyl-maleimide, 1-benzyl-2,5-dihydro-1h-pyrrole-2,5-dione, n-benzylmaleimide dihydrate
PubChem CID 74204
IUPAC Name 1-benzyl-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES O=C1C=CC(=O)N1CC1=CC=CC=C1

Specifications

Melting Point 70°C
Color White
Boiling Point 177°C
Quantity 5 g
UN Number 1759
Formula Weight 187.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylmaleimide
EINECSNumber (5)-6426
TSCA No