missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylglycine Ethyl Ester 96.0+%, TCI America™

$53.90 - $207.92

Chemical Identifiers

CAS 6436-90-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD00009174
InChI Key ULOLIZHBYWAICY-UHFFFAOYSA-N
Synonym n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
PubChem CID 80908
IUPAC Name ethyl 2-(benzylamino)acetate
SMILES CCOC(=O)CNCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22735G
View Documents
TCI America
B22735G
5 g
Each for $53.90
Only null left
 
B227325G
View Documents
TCI America
B227325G
25 g
Each for $207.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6436-90-4
193.25
ULOLIZHBYWAICY-UHFFFAOYSA-N
80908
CCOC(=O)CNCC1=CC=CC=C1
C11H15NO2
MFCD00009174
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
ethyl 2-(benzylamino)acetate

Specifications

6436-90-4
142°C
MFCD00009174
n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
CCOC(=O)CNCC1=CC=CC=C1
193.25
193.25
Liquid
Yellow
C11H15NO2
5 g
ULOLIZHBYWAICY-UHFFFAOYSA-N
ethyl 2-(benzylamino)acetate
80908
≥96.0% (GC)
N-Benzylglycine Ethyl Ester
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1723

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents