missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylglycine, 98+%

Catalog No. AAL1536806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1536806 5 g
AAL1536814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1536806 Supplier Thermo Scientific Chemicals Supplier No. L1536806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17136-36-6 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17136-36-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00190772
InChI Key KGSVNOLLROCJQM-UHFFFAOYSA-N
Synonym n-benzylglycine, 2-benzylamino acetic acid, n-benzyl glycine, glycine, n-phenylmethyl, n-phenylmethyl glycine, benzylglycine, benzylamino acetic acid, n-benzylglycin, bzl-gly-oh
PubChem CID 86965
IUPAC Name 2-(benzylamino)acetic acid
SMILES C1=CC=C(C=C1)CNCC(=O)O

Specifications

Melting Point ∽210°C (decomposition)
Beilstein 2209788
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98%
Chemical Name or Material N-Benzylglycine
EINECSNumber 241-194-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only