CAS RN 17136-36-6
IUPAC Name:
2-(benzylamino)acetic acid
Synonyms:
(benzylamino)acetic acid
Molecular Weight (g/mol):
165.19
Molecular Formula:
C9H11NO2
InChi Key:
KGSVNOLLROCJQM-UHFFFAOYSA-N
SMILES:
OC(=O)CNCC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
N-Benzylglycine, 98+%
N-Benzylglycine, 98+%
CAS: 17136-36-6 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.192 MDL Number: MFCD00190772 InChI Key: KGSVNOLLROCJQM-UHFFFAOYSA-N Synonym: n-benzylglycine,2-benzylamino acetic acid,n-benzyl glycine,glycine, n-phenylmethyl,n-phenylmethyl glycine,benzylglycine,benzylamino acetic acid,n-benzylglycin,bzl-gly-oh PubChem CID: 86965 IUPAC Name: 2-(benzylamino)acetic acid SMILES: C1=CC=C(C=C1)CNCC(=O)O