missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylethanolamine 95.0+%, TCI America™

Catalog No. b1130500ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
B1130500ML 500 mL
B113025ML 25 mL
2 options

Catalog No. B1130500ML

Supplier: TCI America B1130500ML

Only null left

Chemical Identifiers

CAS 104-63-2
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002840
InChI Key XNIOWJUQPMKCIJ-UHFFFAOYSA-N
Synonym n-benzylethanolamine, 2-benzylamino ethanol, benzylaminoethanol, benzylethanolamine, ethanol, 2-phenylmethyl amino, 2-benzylamino ethan-1-ol, benzyl ethanolamine, n-benzyle ethanol amine, ethanol, 2-benzylamino, unii-48121ms9jm
PubChem CID 4348
IUPAC Name 2-(benzylamino)ethan-1-ol
SMILES OCCNCC1=CC=CC=C1

Specifications

pH 11 (50g/L H2O soln. 20°C)
Boiling Point 280°C
Quantity 500 mL
Formula Weight 151.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Benzylethanolamine
EINECSNumber (3)-0314
TSCA Yes