missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylethanolamine 95.0+%, TCI America™

Catalog No. B113025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B113025ML 25 mL
B1130500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B113025ML Supplier TCI America Supplier No. B113025ML

Chemical Identifiers

CAS 104-63-2
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002840
InChI Key XNIOWJUQPMKCIJ-UHFFFAOYSA-N
Synonym n-benzylethanolamine, 2-benzylamino ethanol, benzylaminoethanol, benzylethanolamine, ethanol, 2-phenylmethyl amino, 2-benzylamino ethan-1-ol, benzyl ethanolamine, n-benzyle ethanol amine, ethanol, 2-benzylamino, unii-48121ms9jm
PubChem CID 4348
IUPAC Name 2-(benzylamino)ethan-1-ol
SMILES OCCNCC1=CC=CC=C1

Specifications

pH 11 (50g/L H2O soln. 20°C)
Boiling Point 280°C
Quantity 25 mL
Formula Weight 151.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Benzylethanolamine
EINECSNumber (3)-0314
TSCA Yes