missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyldimethylamine, 98+%

Catalog No. AAA12191AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12191AU 1000 mL
AAA12191AK 250 mL
AAA121910J 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12191AU Supplier Thermo Scientific Chemicals Supplier No. A12191AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-83-3 | C9H13N | 135.21 g/mol

N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylbenzylamine is used in the preparation of bis[(N,N-dimethylamino)benzyl] selenide. It acts as a catalyst in the curing reaction of formulations of diglycidyl ether of bisphenol A and tetrahydrophthalic anhydride. It undergoes directed ortho metalation with butyl lithium. It reacts with methyl iodide to get ammonium salt, which is used as phase transfer catalysts. Further, it is used as a catalyst for the formation of polyurethane foams and epoxy resins.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong acids, strong bases, strong oxidizing agents, strong reducing agents and carbon dioxide.

Chemical Identifiers

CAS 103-83-3
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008329
InChI Key XXBDWLFCJWSEKW-UHFFFAOYSA-N
Synonym n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
PubChem CID 7681
IUPAC Name N,N-dimethyl-1-phenylmethanamine
SMILES CN(C)CC1=CC=CC=C1

Specifications

Melting Point -75°C
Density 0.9
Boiling Point 183°C to 184°C
Flash Point 54°C (129°F)
Refractive Index 1.501
Linear Formula C6H5CH2N(CH3)2
Quantity 1000 mL
UN Number UN2619
Beilstein 1099620
Sensitivity Air sensitive
Solubility Information Slightly miscible with water.
Formula Weight 135.21
Percent Purity ≥98%
Chemical Name or Material N-Benzyldimethylamine
Show More Show Less
Hazard Category H226-H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZYLDIMETHYLAMINE
EINECSNumber 203-149-1
RTECSNumber DP4500000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only