missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylacrylamide, 96%

Catalog No. AAL1145003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1145003 1 g
AAL1145006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1145003 Supplier Thermo Scientific Chemicals Supplier No. L1145003

CAS: 13304-62-6 | C10H11NO | 161.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13304-62-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00015333
InChI Key OHLHOLGYGRKZMU-UHFFFAOYSA-N
Synonym n-benzylacrylamide, 2-propenamide, n-phenylmethyl, n-benzyl acrylamide, acmc-20ak6s, enamine_005841
PubChem CID 139428
IUPAC Name N-benzylprop-2-enamide
SMILES C=CC(=O)NCC1=CC=CC=C1

Specifications

Melting Point 59°C to 62°C
Quantity 1 g
Beilstein 2690053
Formula Weight 161.21
Percent Purity 96%
Chemical Name or Material N-Benzylacrylamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 603-701-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only