missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-tert-butylamine, 96%

CAS: 3378-72-1 | C11H17N | 163.264 g/mol

$156.40 - $423.67

Chemical Identifiers

CAS 3378-72-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00008798
InChI Key DLSOILHAKCBARI-UHFFFAOYSA-N
Synonym n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
PubChem CID 76908
IUPAC Name N-benzyl-2-methylpropan-2-amine
SMILES CC(C)(C)NCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0114614
View Documents
Thermo Scientific Chemicals
L0114614
25 g
Each for $156.40
Only null left
 
AAL0114622
View Documents
Thermo Scientific Chemicals
L0114622
100 g
Each for $423.67
Only null left
 
Description

Description

N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

3378-72-1
163.264
DLSOILHAKCBARI-UHFFFAOYSA-N
76908
CC(C)(C)NCC1=CC=CC=C1
C11H17N
MFCD00008798
n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
N-benzyl-2-methylpropan-2-amine

Specifications

3378-72-1
0.899
88°C (190°F)
1.497
MFCD00008798
UN2735
Air Sensitive
Insoluble in water.
CC(C)(C)NCC1=CC=CC=C1
163.264
163.27
N-Benzyl-tert-butylamine
-30°C to -25°C
218°C to 220°C
C11H17N
C6H5CH2NHC(CH3)3
25 g
507464
n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
DLSOILHAKCBARI-UHFFFAOYSA-N
N-benzyl-2-methylpropan-2-amine
76908
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 222-179-6

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only