missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-tert-butylamine, 96%

Catalog No. AAL0114622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0114622 100 g
AAL0114614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0114622 Supplier Thermo Scientific Chemicals Supplier No. L0114622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3378-72-1 | C11H17N | 163.264 g/mol

N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 3378-72-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00008798
InChI Key DLSOILHAKCBARI-UHFFFAOYSA-N
Synonym n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
PubChem CID 76908
IUPAC Name N-benzyl-2-methylpropan-2-amine
SMILES CC(C)(C)NCC1=CC=CC=C1

Specifications

Melting Point -30°C to -25°C
Density 0.899
Boiling Point 218°C to 220°C
Flash Point 88°C (190°F)
Refractive Index 1.497
Linear Formula C6H5CH2NHC(CH3)3
Quantity 100 g
UN Number UN2735
Beilstein 507464
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 163.27
Percent Purity 96%
Chemical Name or Material N-Benzyl-tert-butylamine
Show More Show Less
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 222-179-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only