missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-p-toluenesulfonamide 98.0+%, TCI America™

Catalog No. B30825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30825G 5 g
B308225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30825G Supplier TCI America Supplier No. B30825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1576-37-0
Molecular Formula C14H15NO2S
Molecular Weight (g/mol) 261.339
MDL Number MFCD00159328
InChI Key WTHKAJZQYNKTCJ-UHFFFAOYSA-N
Synonym n-benzyl-p-toluenesulfonamide, n-tosylbenzylamine, p-toluenesulfonamide, n-benzyl, n-benzyl-p-toluenesulphonamide, n-benzyl-4-methyl-benzenesulfonamide, n-benzyl-4-methylbenzene-1-sulfonamide, n-benzyl-p-toluene sulphonamide, benzenesulfonamide, 4-methyl-n-phenylmethyl, 4-methyl-n-phenylmethyl benzenesulfonamide, 4-methyl-n-phenylmethyl-benzenesulfonamid
PubChem CID 95801
IUPAC Name N-benzyl-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NCC2=CC=CC=C2

Specifications

Melting Point 115°C
Color White
Quantity 5 g
Formula Weight 261.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-p-toluenesulfonamide
RTECSNumber XT5490000
TSCA No