missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyl-p-anisidine 98.0+%, TCI America™
$136.08 - $136.08
Chemical Identifiers
| CAS | 17377-95-6 |
|---|---|
| Molecular Formula | C14H15NO |
| Molecular Weight (g/mol) | 213.28 |
| MDL Number | MFCD00059298 |
| InChI Key | LIJJGMDKVVOEFT-UHFFFAOYSA-N |
| Synonym | n-benzyl-p-anisidine, n-4-methoxyphenyl-n-benzylamine, 4-methoxyphenyl benzylamine, n-benzyl-para-anisidine, acmc-1bvsm, n-benzyl-p-methoxyaniline, cambridge id 5107549, n-benzyl-4-methoxy-aniline, n-4-methoxyphenyl benzylamine, n-4-methoxyphenyl benzyl amine |
| PubChem CID | 519413 |
| IUPAC Name | N-benzyl-4-methoxyaniline |
| SMILES | COC1=CC=C(NCC2=CC=CC=C2)C=C1 |
Chemical Identifiers
| 17377-95-6 | |
| 213.28 | |
| LIJJGMDKVVOEFT-UHFFFAOYSA-N | |
| 519413 | |
| COC1=CC=C(NCC2=CC=CC=C2)C=C1 |
| C14H15NO | |
| MFCD00059298 | |
| n-benzyl-p-anisidine, n-4-methoxyphenyl-n-benzylamine, 4-methoxyphenyl benzylamine, n-benzyl-para-anisidine, acmc-1bvsm, n-benzyl-p-methoxyaniline, cambridge id 5107549, n-benzyl-4-methoxy-aniline, n-4-methoxyphenyl benzylamine, n-4-methoxyphenyl benzyl amine | |
| N-benzyl-4-methoxyaniline |
Specifications
| 17377-95-6 | |
| Yellow | |
| C14H15NO | |
| 5 g | |
| LIJJGMDKVVOEFT-UHFFFAOYSA-N | |
| N-benzyl-4-methoxyaniline | |
| 519413 | |
| ≥98.0% (GC,T) | |
| N-Benzyl-p-anisidine |
| 48°C | |
| 238°C | |
| MFCD00059298 | |
| n-benzyl-p-anisidine, n-4-methoxyphenyl-n-benzylamine, 4-methoxyphenyl benzylamine, n-benzyl-para-anisidine, acmc-1bvsm, n-benzyl-p-methoxyaniline, cambridge id 5107549, n-benzyl-4-methoxy-aniline, n-4-methoxyphenyl benzylamine, n-4-methoxyphenyl benzyl amine | |
| COC1=CC=C(NCC2=CC=CC=C2)C=C1 | |
| 213.28 | |
| 213.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No