Learn More
N-Benzyl-N-phenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals
$88.45 - $298.27
Chemical Identifiers
CAS | 5705-15-7 |
---|---|
Molecular Formula | C13H15ClN2 |
Molecular Weight (g/mol) | 234.73 |
MDL Number | MFCD00050690 |
InChI Key | JTYLHYOCBGPMNO-UHFFFAOYSA-N |
Synonym | 1-benzyl-1-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazine hydrochloride, usaf ek-1666, hydrazine, 1-phenyl-1-phenylmethyl-, monohydrochloride, alpha-benzyl-alpha-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazinium 1+ chloride, hydrazine, 1-benzyl-1-phenyl-, hydrochloride, 1-benzyl-1-phenylhydrazine hydrochloride van, hydrazine, 1-phenyl-1-phenylmethyl-, hydrochloride 1:1, acmc-209lvq |
PubChem CID | 79785 |
IUPAC Name | 1-benzyl-1-phenylhydrazine;hydrochloride |
SMILES | [H+].[Cl-].NN(CC1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1736706
|
Thermo Scientific Chemicals
A1736706 |
5 g |
Each for $88.45
|
|
|||||
AAA1736714
|
Thermo Scientific Chemicals
A1736714 |
25 g |
Each for $298.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5705-15-7 | |
234.73 | |
JTYLHYOCBGPMNO-UHFFFAOYSA-N | |
79785 | |
[H+].[Cl-].NN(CC1=CC=CC=C1)C1=CC=CC=C1 |
C13H15ClN2 | |
MFCD00050690 | |
1-benzyl-1-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazine hydrochloride, usaf ek-1666, hydrazine, 1-phenyl-1-phenylmethyl-, monohydrochloride, alpha-benzyl-alpha-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazinium 1+ chloride, hydrazine, 1-benzyl-1-phenyl-, hydrochloride, 1-benzyl-1-phenylhydrazine hydrochloride van, hydrazine, 1-phenyl-1-phenylmethyl-, hydrochloride 1:1, acmc-209lvq | |
1-benzyl-1-phenylhydrazine;hydrochloride |
Specifications
5705-15-7 | |
C13H15ClN2 | |
5 g | |
1-benzyl-1-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazine hydrochloride, usaf ek-1666, hydrazine, 1-phenyl-1-phenylmethyl-, monohydrochloride, alpha-benzyl-alpha-phenylhydrazine hydrochloride, n-benzyl-n-phenylhydrazinium 1+ chloride, hydrazine, 1-benzyl-1-phenyl-, hydrochloride, 1-benzyl-1-phenylhydrazine hydrochloride van, hydrazine, 1-phenyl-1-phenylmethyl-, hydrochloride 1:1, acmc-209lvq | |
[H+].[Cl-].NN(CC1=CC=CC=C1)C1=CC=CC=C1 | |
234.73 | |
234.73 | |
N-Benzyl-N-phenylhydrazine hydrochloride |
178°C to 181°C | |
MFCD00050690 | |
3707186 | |
JTYLHYOCBGPMNO-UHFFFAOYSA-N | |
1-benzyl-1-phenylhydrazine;hydrochloride | |
79785 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 227-195-7
RTECSNumber : MU9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only