missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-N-nitroso-p-toluenesulfonamide 98.0+%, TCI America™

Catalog No. p-7119918
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B30515G 5 g
B305125G 25 g
2 options

Catalog No. B30515G

Supplier: TCI America B30515G

Only null left

Chemical Identifiers

CAS 33528-13-1
Molecular Formula C14H14N2O3S
Molecular Weight (g/mol) 290.337
MDL Number MFCD00216617
InChI Key ZXELPGWSCCGNDS-UHFFFAOYSA-N
PubChem CID 4118076
IUPAC Name N-benzyl-4-methyl-N-nitrosobenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(CC2=CC=CC=C2)N=O

Specifications

Melting Point 85°C
Color Yellow
Quantity 5 g
Formula Weight 290.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-N-nitroso-p-toluenesulfonamide
TSCA No
Recommended Storage Refrigerator