missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[(N-Benzyl-N-methylammonium)methyl]trifluoroborate internal salt, 95 %, Thermo Scientific™
Chemical Identifiers
| CAS | 1268341-00-9 |
|---|---|
| Molecular Formula | C9H13BF3N |
| Molecular Weight (g/mol) | 203.015 |
| MDL Number | MFCD20134169 |
| InChI Key | JRTRNQRZOUGDCI-UHFFFAOYSA-O |
| Synonym | n-benzyl-n-methylammonium methyl trifluoroborate internal salt, benzyl methyl ammonio methyl trifluoroboranuide |
| PubChem CID | 53243645 |
| IUPAC Name | [benzyl(methyl)azaniumyl]methyl-trifluoroboranuide |
| SMILES | [B-](C[NH+](C)CC1=CC=CC=C1)(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH3339703
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1268341-00-9 | |
| 203.015 | |
| JRTRNQRZOUGDCI-UHFFFAOYSA-O | |
| 53243645 | |
| [B-](C[NH+](C)CC1=CC=CC=C1)(F)(F)F |
| C9H13BF3N | |
| MFCD20134169 | |
| n-benzyl-n-methylammonium methyl trifluoroborate internal salt, benzyl methyl ammonio methyl trifluoroboranuide | |
| [benzyl(methyl)azaniumyl]methyl-trifluoroboranuide |
Specifications
| 1268341-00-9 | |
| MFCD20134169 | |
| JRTRNQRZOUGDCI-UHFFFAOYSA-O | |
| [benzyl(methyl)azaniumyl]methyl-trifluoroboranuide | |
| 53243645 | |
| 95% |
| C9H13BF3N | |
| n-benzyl-n-methylammonium methyl trifluoroborate internal salt, benzyl methyl ammonio methyl trifluoroboranuide | |
| [B-](C[NH+](C)CC1=CC=CC=C1)(F)(F)F | |
| 203.015 | |
| 203.02 | |
| [(N-Benzyl-N-methylammonium)methyl]trifluoroborate internal salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only