CAS RN 1268341-00-9
IUPAC Name:
{[benzyl(methyl)azaniumyl]methyl}trifluoroboranuide
Synonyms:
{[benzyl(methyl)ammonio]methyl}trifluoroboranuide
Molecular Weight (g/mol):
203.02
Molecular Formula:
C9H13BF3N
InChi Key:
JRTRNQRZOUGDCI-UHFFFAOYNA-O
SMILES:
C[NH+](CC1=CC=CC=C1)C[B-](F)(F)F
1
–
1
of
1
results
1
Promotions Available
[(N-Benzyl-N-methylammonium)methyl]trifluoroborate internal salt, 95 %, Thermo Scientific™
[(N-Benzyl-N-methylammonium)methyl]trifluoroborate internal salt, 95 %, Thermo Scientific™
CAS: 1268341-00-9 Molecular Formula: C9H13BF3N Molecular Weight (g/mol): 203.015 MDL Number: MFCD20134169 InChI Key: JRTRNQRZOUGDCI-UHFFFAOYSA-O Synonym: n-benzyl-n-methylammonium methyl trifluoroborate internal salt,benzyl methyl ammonio methyl trifluoroboranuide PubChem CID: 53243645 IUPAC Name: [benzyl(methyl)azaniumyl]methyl-trifluoroboranuide SMILES: [B-](C[NH+](C)CC1=CC=CC=C1)(F)(F)F