missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-N-(methoxymethyl)-N-trimethylsilylmethylamine 98.0+%, TCI America™

Supplier:  TCI America B193825G

 View more versions of this product

Catalog No. B193825G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Benzyl-N-(methoxymethyl)-N-trimethylsilylmethylamine
Yellow
C13H23NOSi
25 g
RPZAAFUKDPKTKP-UHFFFAOYSA-N
N-(methoxymethyl)-1-phenyl-N-(trimethylsilylmethyl)methanamine
353442
≥98.0% (T)
93102-05-7
78°C
MFCD00674005
n-methoxymethyl-n-trimethylsilylmethyl benzylamine, n-benzyl-1-methoxy-n-trimethylsilyl methyl methanamine, benzyl methoxymethyl trimethylsilyl methyl amine, benzyl-methoxymethyl-trimethylsilanylmethyl-amine, n-benzyl-n-methoxymethyl-n-trimethylsilylmethylamine, n-benzyl-n-methoxymethyl trimethylsilylmethylamine, n-methoxymethyl phenyl-n-trimethylsilyl methyl methanamine, n-methoxymethyl-n-trimethylsilyl methyl phenyl, n-methoxymethyl-n-trimethylsilylmethyl-benzylamine
COCN(CC1=CC=CC=C1)C[Si](C)(C)C
237.418
237.42
Liquid

Chemical Identifiers

93102-05-7
237.418
RPZAAFUKDPKTKP-UHFFFAOYSA-N
353442
COCN(CC1=CC=CC=C1)C[Si](C)(C)C
C13H23NOSi
MFCD00674005
n-methoxymethyl-n-trimethylsilylmethyl benzylamine, n-benzyl-1-methoxy-n-trimethylsilyl methyl methanamine, benzyl methoxymethyl trimethylsilyl methyl amine, benzyl-methoxymethyl-trimethylsilanylmethyl-amine, n-benzyl-n-methoxymethyl-n-trimethylsilylmethylamine, n-benzyl-n-methoxymethyl trimethylsilylmethylamine, n-methoxymethyl phenyl-n-trimethylsilyl methyl methanamine, n-methoxymethyl-n-trimethylsilyl methyl phenyl, n-methoxymethyl-n-trimethylsilylmethyl-benzylamine
N-(methoxymethyl)-1-phenyl-N-(trimethylsilylmethyl)methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions