missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-N-(methoxymethyl)-N-trimethylsilylmethylamine 98.0+%, TCI America™

Catalog No. p-7119367
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19385G 5 g
B193825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19385G Supplier TCI America Supplier No. B19385G
Only null left

Chemical Identifiers

CAS 93102-05-7
Molecular Formula C13H23NOSi
Molecular Weight (g/mol) 237.418
MDL Number MFCD00674005
InChI Key RPZAAFUKDPKTKP-UHFFFAOYSA-N
Synonym n-methoxymethyl-n-trimethylsilylmethyl benzylamine, n-benzyl-1-methoxy-n-trimethylsilyl methyl methanamine, benzyl methoxymethyl trimethylsilyl methyl amine, benzyl-methoxymethyl-trimethylsilanylmethyl-amine, n-benzyl-n-methoxymethyl-n-trimethylsilylmethylamine, n-benzyl-n-methoxymethyl trimethylsilylmethylamine, n-methoxymethyl phenyl-n-trimethylsilyl methyl methanamine, n-methoxymethyl-n-trimethylsilyl methyl phenyl, n-methoxymethyl-n-trimethylsilylmethyl-benzylamine
PubChem CID 353442
IUPAC Name N-(methoxymethyl)-1-phenyl-N-(trimethylsilylmethyl)methanamine
SMILES COCN(CC1=CC=CC=C1)C[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
Formula Weight 237.42
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N-Benzyl-N-(methoxymethyl)-N-trimethylsilylmethylamine
TSCA No
Recommended Storage Refrigerator