missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-methylbenzamide, 97%, Thermo Scientific™

$149.70 - $149.70

Chemical Identifiers

CAS 5436-83-9
Molecular Formula C15H15NO
Molecular Weight (g/mol) 225.291
MDL Number MFCD00026008
InChI Key QIULRTIURUDCGH-UHFFFAOYSA-N
Synonym benzamide, 4-methyl-n-phenylmethyl, cambridge id 5104261, benzamide,4-methyl-n-phenylmethyl
PubChem CID 228681
IUPAC Name N-benzyl-4-methylbenzamide
SMILES CC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH56270MD
View Documents
Thermo Scientific Chemicals
H56270MD
250 mg
Each for $149.70
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. 

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

5436-83-9
225.291
QIULRTIURUDCGH-UHFFFAOYSA-N
228681
CC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2
C15H15NO
MFCD00026008
benzamide, 4-methyl-n-phenylmethyl, cambridge id 5104261, benzamide,4-methyl-n-phenylmethyl
N-benzyl-4-methylbenzamide

Specifications

5436-83-9
MFCD00026008
benzamide, 4-methyl-n-phenylmethyl, cambridge id 5104261, benzamide,4-methyl-n-phenylmethyl
CC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2
225.291
225.29
N-Benzyl-4-methylbenzamide
C15H15NO
250 mg
QIULRTIURUDCGH-UHFFFAOYSA-N
N-benzyl-4-methylbenzamide
228681
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'