missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2,2,2-trifluoroacetamide 95.0+%, TCI America™

Catalog No. b57055g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B57055G 5 g
B57051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B57055G Supplier TCI America Supplier No. B57055G

Chemical Identifiers

CAS 7387-69-1
Molecular Formula C9H8F3NO
Molecular Weight (g/mol) 203.164
MDL Number MFCD00188542
InChI Key DEXVYKWXVWAYGO-UHFFFAOYSA-N
PubChem CID 230121
IUPAC Name N-benzyl-2,2,2-trifluoroacetamide
SMILES C1=CC=C(C=C1)CNC(=O)C(F)(F)F

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 5 g
Formula Weight 203.16
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-2,2,2-trifluoroacetamide