accessibility menu, dialog, popup

UserName

FFFOHN

CAS RN 7387-69-1

IUPAC Name: N-benzyl-2,2,2-trifluoroacetamide
Synonyms: N-benzyl-2,2,2-trifluoroacetamide
Molecular Weight (g/mol): 203.16
Molecular Formula: C9H8F3NO
InChi Key: DEXVYKWXVWAYGO-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)NCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results