missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoylthiourea, 98%

Catalog No. AAL1237714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1237714 25 g
AAL1237706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1237714 Supplier Thermo Scientific Chemicals Supplier No. L1237714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 614-23-3 | C8H8N2OS | 180.225 g/mol

N-Benzoylthiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoylthiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 614-23-3
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.225
MDL Number MFCD00041191
InChI Key DQMWMUMCNOJLSI-UHFFFAOYSA-N
Synonym benzoylthiourea, n-benzoylthiourea, 1-benzoyl-2-thiourea, necacyl, 1-benzoylthiourea, benzamide, n-thiocarbamoyl, urea, 1-benzoyl-2-thio, benzamide, n-aminothioxomethyl, n-aminothioxomethyl benzamide, benzamide, n-aminothioxomethyl-9ci
PubChem CID 2735473
IUPAC Name N-carbamothioylbenzamide
SMILES C1=CC=C(C=C1)C(=O)NC(=S)N

Specifications

Melting Point 174°C to 176°C
Odor Odorless
Quantity 25 g
Beilstein 1942772
Solubility Information Insoluble in water.
Formula Weight 180.23
Percent Purity 98%
Chemical Name or Material N-Benzoylthiourea
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber YS1450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only