missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoylaminopurine, 99%

Catalog No. AAL0829214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0829214 25 g
AAL0829203 1 g
AAL0829206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0829214 Supplier Thermo Scientific Chemicals Supplier No. L0829214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4005-49-6 | C12H9N5O | 239.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4005-49-6
Molecular Formula C12H9N5O
Molecular Weight (g/mol) 239.238
MDL Number MFCD00037927
InChI Key QQJXZVKXNSFHRI-UHFFFAOYSA-N
Synonym n6-benzoyladenine, n-7h-purin-6-yl benzamide, n-benzoylaminopurine, n-9h-purin-6-yl benzamide, 6-benzoylaminopurine, benzamide, n-1h-purin-6-yl, 6-benzamidopurine, adenine,6n-benzoyl, phenyl-n-purin-6-ylcarboxamide, n-benzoyladenine
PubChem CID 97075
IUPAC Name N-(7H-purin-6-yl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=NC=NC3=C2NC=N3

Specifications

Melting Point 241°C to 244°C
Quantity 25 g
Beilstein 20585
Formula Weight 239.23
Percent Purity 99%
Chemical Name or Material N-Benzoylaminopurine
Hazard Category H302-H317
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only