missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoyl-N-phenylhydroxylamine 98.0+%, TCI America™
Supplier: TCI America P015825G
Specifications
| N-Benzoyl-N-phenylhydroxylamine | |
| 122°C | |
| C13H11NO2 | |
| 25 g | |
| YLYIXDZITBMCIW-UHFFFAOYSA-N | |
| N-hydroxy-N-phenylbenzamide | |
| 67536 | |
| ≥98.0% (N) |
| 304-88-1 | |
| White | |
| MFCD00002111 | |
| n-benzoyl-n-phenylhydroxylamine, n-phenylbenzohydroxamic acid, n-hydroxybenzanilide, benzamide, n-hydroxy-n-phenyl, n-benzoyl-n-phenylhydroxamic acid, n-benzoylphenylhydroxylamine, benzohydroxamic acid, n-phenyl, tantalon, acmc-1cqm9, n-phenylbenzohydroxamicacid | |
| ON(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 213.24 | |
| 213.24 | |
| Crystalline Powder |
Chemical Identifiers
| 304-88-1 | |
| 213.24 | |
| YLYIXDZITBMCIW-UHFFFAOYSA-N | |
| 67536 | |
| ON(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
| C13H11NO2 | |
| MFCD00002111 | |
| n-benzoyl-n-phenylhydroxylamine, n-phenylbenzohydroxamic acid, n-hydroxybenzanilide, benzamide, n-hydroxy-n-phenyl, n-benzoyl-n-phenylhydroxamic acid, n-benzoylphenylhydroxylamine, benzohydroxamic acid, n-phenyl, tantalon, acmc-1cqm9, n-phenylbenzohydroxamicacid | |
| N-hydroxy-N-phenylbenzamide |
Safety and Handling
TSCA : Yes