missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-N-phenylhydroxylamine, 98%

Catalog No. AAA1195006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1195006 5 g
AAA1195014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1195006 Supplier Thermo Scientific Chemicals Supplier No. A1195006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 304-88-1 | C13H11NO2 | 213.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 304-88-1
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.24
MDL Number MFCD00002111
InChI Key YLYIXDZITBMCIW-UHFFFAOYSA-N
Synonym n-benzoyl-n-phenylhydroxylamine, n-phenylbenzohydroxamic acid, n-hydroxybenzanilide, benzamide, n-hydroxy-n-phenyl, n-benzoyl-n-phenylhydroxamic acid, n-benzoylphenylhydroxylamine, benzohydroxamic acid, n-phenyl, tantalon, acmc-1cqm9, n-phenylbenzohydroxamicacid
PubChem CID 67536
IUPAC Name N-hydroxy-N-phenylbenzamide
SMILES ON(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 118°C to 120°C
Quantity 5 g
Beilstein 2212449
Formula Weight 213.24
Percent Purity 98%
Chemical Name or Material N-Benzoyl-N-phenylhydroxylamine
EINECSNumber 206-158-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only