missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoyl-L-tyrosine 98.0+%, TCI America™
$249.78 - $756.81
Chemical Identifiers
| CAS | 2566-23-6 |
|---|---|
| Molecular Formula | C16H15NO4 |
| Molecular Weight (g/mol) | 285.299 |
| MDL Number | MFCD01321140 |
| InChI Key | KUUUDPTUEOKITK-AWEZNQCLSA-N |
| PubChem CID | 151021 |
| IUPAC Name | (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoic acid |
| SMILES | C1=CC=C(C=C1)C(=O)NC(CC2=CC=C(C=C2)O)C(=O)O |
Chemical Identifiers
| 2566-23-6 | |
| 285.299 | |
| KUUUDPTUEOKITK-AWEZNQCLSA-N | |
| (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoic acid |
| C16H15NO4 | |
| MFCD01321140 | |
| 151021 | |
| C1=CC=C(C=C1)C(=O)NC(CC2=CC=C(C=C2)O)C(=O)O |
Specifications
| 2566-23-6 | |
| C16H15NO4 | |
| 5 g | |
| C1=CC=C(C=C1)C(=O)NC(CC2=CC=C(C=C2)O)C(=O)O | |
| 285.299 | |
| 285.30 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01321140 | |
| KUUUDPTUEOKITK-AWEZNQCLSA-N | |
| (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoic acid | |
| 151021 | |
| ≥98.0% (HPLC,T) | |
| N-Benzoyl-L-tyrosine |
Safety and Handling
TSCA : No