accessibility menu, dialog, popup

UserName

CAS RN 2566-23-6

OHO(S)HONHO

CAS RN 2566-23-6

IUPAC Name: (2S)-3-(4-hydroxyphenyl)-2-(phenylformamido)propanoic acid
Synonyms: (2S)-3-(4-hydroxyphenyl)-2-(phenylformamido)propanoic acid
Molecular Weight (g/mol): 285.3
Molecular Formula: C16H15NO4
InChi Key: KUUUDPTUEOKITK-AWEZNQCLSA-N
SMILES: OC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results