missing translation for 'onlineSavingsMsg'
Learn More

N-α-Benzoyl-L-Arginine p-Nitroanilide Hydrochloride, MP Biomedicals™

Catalog No. icn15482980
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15482980 100 mg
ICN15482925 25 mg
ICN15482983 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15482980 Supplier MP Biomedicals Inc Supplier No. 0215482980

Chemical Identifiers

CAS 21653-40-7
Molecular Formula C19H23ClN6O4
Molecular Weight (g/mol) 434.881
MDL Number MFCD00063682
InChI Key DEOKFPFLXFNAON-NTISSMGPSA-N
Synonym bz-arg-pna.hcl, s-n-5-guanidino-1-4-nitrophenyl amino-1-oxopentan-2-yl benzamide hydrochloride, nalpha-benzoyl-l-arginine 4-nitroanilide hydrochloride, bani, s-5-amidinoamino-2-benzoylamino-n-4-nitrophenyl valeramide monohydrochloride, l-bapna, 2s-5-carbamimidamido-n-4-nitrophenyl-2-phenylformamido pentanamide hydrochloride, bz-arg-pna-hcl, bz-arg-pna hcl, bz-arg-pna . hcl
PubChem CID 16219022
IUPAC Name N-[(2S)-5-(diaminomethylideneamino)-1-(4-nitroanilino)-1-oxopentan-2-yl]benzamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)NC2=CC=C(C=C2)[N+](=O)[O-].Cl

Specifications

Color White
Assay Percent Range >99%
Quantity 100 mg
Formula Weight 434.9
Physical Form Powder
Chemical Name or Material N-α-Benzoyl-L-Arginine p-Nitroanilide Hydrochloride
EINECSNumber 244-505-6
Recommended Storage Store at -0°C.