missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoyl-DL-phenylalanine 2-Naphthyl Ester 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 158°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 395.46 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Benzoyl-DL-phenylalanine 2-Naphthyl Ester [for Determination of Chymotrypsin] |
Safety and Handling
| TSCA | No |