accessibility menu, dialog, popup

UserName

OONHO

CAS RN 15873-25-3

IUPAC Name: naphthalen-2-yl 3-phenyl-2-(phenylformamido)propanoate
Synonyms: naphthalen-2-yl 3-phenyl-2-(phenylformamido)propanoate
Molecular Weight (g/mol): 395.46
Molecular Formula: C26H21NO3
InChi Key: NPPKNSHRAVHLHD-UHFFFAOYNA-N
SMILES: O=C(OC1=CC=C2C=CC=CC2=C1)C(CC1=CC=CC=C1)NC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results