missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-DL-phenylalanine 2-Naphthyl Ester 98.0+%, TCI America™

Catalog No. B0309100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0309100MG 100 mg
B03091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0309100MG Supplier TCI America Supplier No. B0309100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15873-25-3
Molecular Formula C26H21NO3
Molecular Weight (g/mol) 395.46
MDL Number MFCD00021607
InChI Key NPPKNSHRAVHLHD-UHFFFAOYNA-N
PubChem CID 102757
IUPAC Name naphthalen-2-yl 3-phenyl-2-(phenylformamido)propanoate
SMILES O=C(OC1=CC=C2C=CC=CC2=C1)C(CC1=CC=CC=C1)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 158°C
Color White
Quantity 100 mg
Formula Weight 395.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzoyl-DL-phenylalanine 2-Naphthyl Ester [for Determination of Chymotrypsin]