missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoyl-DL-alanine, 97+%
CAS: 1205-02-3 | C10H11NO3 | 193.20 g/mol
$73.64 - $238.51
Chemical Identifiers
| CAS | 1205-02-3 |
|---|---|
| Molecular Formula | C10H11NO3 |
| Molecular Weight (g/mol) | 193.20 |
| MDL Number | MFCD00020393 |
| InChI Key | UAQVHNZEONHPQG-UHFFFAOYNA-N |
| Synonym | n-benzoyl-dl-alanine, benzoyl-dl-alanine, methylhippuric acid, n-benzoylalanine, alanine, n-benzoyl, benzoylalanine, dl-n-benzoylalanine, 2-phenylformamido propanoic acid, alpha-methylhippuric acid, n-phenylcarbonyl alanine |
| PubChem CID | 71002 |
| ChEBI | CHEBI:71167 |
| IUPAC Name | 2-benzamidopropanoic acid |
| SMILES | CC(NC(=O)C1=CC=CC=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1310009
|
Thermo Scientific Chemicals
L1310009 |
10 g |
Each for $73.64
|
|
|||||
|
AAL1310018
|
Thermo Scientific Chemicals
L1310018 |
50 g |
Each for $238.51
|
|
|||||
Description
N-Benzoyl-DL-alanine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Benzoyl-DL-alanine is used as pharmaceutical intermediate.
Solubility
Very slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 1205-02-3 | |
| 193.20 | |
| UAQVHNZEONHPQG-UHFFFAOYNA-N | |
| 71002 | |
| 2-benzamidopropanoic acid |
| C10H11NO3 | |
| MFCD00020393 | |
| n-benzoyl-dl-alanine, benzoyl-dl-alanine, methylhippuric acid, n-benzoylalanine, alanine, n-benzoyl, benzoylalanine, dl-n-benzoylalanine, 2-phenylformamido propanoic acid, alpha-methylhippuric acid, n-phenylcarbonyl alanine | |
| CHEBI:71167 | |
| CC(NC(=O)C1=CC=CC=C1)C(O)=O |
Specifications
| 163°C to 167°C | |
| C10H11NO3 | |
| 3201778 | |
| Very slightly soluble in water. | |
| CC(NC(=O)C1=CC=CC=C1)C(O)=O | |
| 10 g | |
| 71002 | |
| 193.2 | |
| N-Benzoyl-DL-alanine |
| 1205-02-3 | |
| MFCD00020393 | |
| n-benzoyl-dl-alanine, benzoyl-dl-alanine, methylhippuric acid, n-benzoylalanine, alanine, n-benzoyl, benzoylalanine, dl-n-benzoylalanine, 2-phenylformamido propanoic acid, alpha-methylhippuric acid, n-phenylcarbonyl alanine | |
| UAQVHNZEONHPQG-UHFFFAOYNA-N | |
| 2-benzamidopropanoic acid | |
| 193.20 | |
| CHEBI:71167 | |
| ≥97% |
Safety and Handling
EINECSNumber : 214-879-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only