missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%

Catalog No. AAH5230903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5230903 1 g
AAH5230906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5230903 Supplier Thermo Scientific Chemicals Supplier No. H5230903
Only null left

CAS: 51549-39-4 | C23H31N5O4Si | 469.62 g/mol

N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoyl-5′-O-tert-butyldimethylsilyl-2′-deoxyadenosine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 51549-39-4
Molecular Formula C23H31N5O4Si
Molecular Weight (g/mol) 469.62
MDL Number MFCD04972281
InChI Key BJZZQJFYKQPUGO-TWAUYLRJNA-N
IUPAC Name N-{9-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide
SMILES CC(C)(C)[Si](C)(C)OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12

Specifications

Quantity 1 g
Formula Weight 469.62
Percent Purity ≥98%
Chemical Name or Material N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only