missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine, 97+%

Catalog No. p-7034617
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5230603 1 g
AAH5230606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5230603 Supplier Thermo Scientific Chemicals Supplier No. H5230603
Only null left

CAS: 140712-79-4 | C38H35N5O6 | 657.73 g/mol

N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoyl-3′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 140712-79-4
Molecular Formula C38H35N5O6
Molecular Weight (g/mol) 657.73
MDL Number MFCD04972282
InChI Key LFXBQKFIXWICJR-BLBFAAIZNA-N
Synonym n6-benzoyl-3'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, adenosine,n-benzoyl-3'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-9ci, n-benzoyl-3'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, n6-bezoyl-3'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, n-benzoyl-3'-o-bis 4-methoxyphenyl phenyl methyl-2'-deoxyadenosine, n-9-2r,4s,5r-4-bis 4-methoxyphenyl phenyl methoxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-yl benzamide, n-9-2r,4s,5r-4-bis 4-methoxyphenyl phenyl methoxy-5-hydroxymethyl oxolan-2-yl purin-6-yl benzamide
PubChem CID 15928822
IUPAC Name N-[9-[(2R,4S,5R)-4-[bis(4-methoxyphenyl)-phenylmethoxy]-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
SMILES COC1=CC=C(C=C1)C(O[C@H]1C[C@@H](O[C@@H]1CO)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Quantity 1 g
Formula Weight 657.73
Percent Purity ≥97%
Chemical Name or Material N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only