missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-2'-deoxycytidine, 98+%

Catalog No. p-7076531
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5229803 1 g
AAH5229806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5229803 Supplier Thermo Scientific Chemicals Supplier No. H5229803
Only null left

CAS: 4836-13-9 | C16H17N3O5 | 331.33 g/mol

Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine, N-Protected 2?-Deoxyribonucleosides(useful for oligonucleotide synthesis), 3?-, 5?-and 3?, 5?-di-O-crotonyl 2?-deoxynucleoside derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4836-13-9
Molecular Formula C16H17N3O5
Molecular Weight (g/mol) 331.33
MDL Number MFCD00010115
InChI Key MPSJHJFNKMUKCN-OUCADQQQSA-N
Synonym n4-benzoyl-2'-deoxycytidine, n-benzoyl-2'-deoxy-cytidine, n-benzoyl-2'-deoxycytidine, cytidine, n-benzoyl-2'-deoxy, cytidine,n-benzoyl-2'-deoxy, n-benzoyl-2-deoxy-cytidine, dc-bz, ksc590o7t, n-benzoyl-2/'-deoxycytidine
PubChem CID 9797617
IUPAC Name N-[1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O

Specifications

Packaging Glass bottle
Quantity 1 g
Solubility Information Soluble in methanol (50mg/ml).
Formula Weight 331.33
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material N-Benzoyl-2′-deoxycytidine
EINECSNumber 225-421-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only