Learn More
N-Arachidonylglycine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J67492LB0
| Quantity | 5 mg |
|---|
Description
An endogenous anandamide-like compound; also acts as a T-type Ca2+ channel blocker and an endogenous GLYT2 inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 179113-91-8 | |
| 361.526 | |
| YLEARPUNMCCKMP-DOFZRALJSA-N | |
| 5283389 | |
| 2-[[(5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoyl]amino]acetic acid |
| C22H35NO3 | |
| MFCD03791313 | |
| n-arachidonylglycine, n-arachidonoylglycine, nagly, n-arachidonoyl glycine, arachidonoyl glycine, n-5z,8z,11z,14z-eicosatetraenoyl-glycine, dsstox_cid_25124, dsstox_rid_80688, dsstox_gsid_45124, 2-5z,8z,11z,14z-icosa-5,8,11,14-tetraenoyl amino acetic acid | |
| CHEBI:58961 | |
| CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCC(=O)O |
Specifications
| N-Arachidonylglycine | |
| White | |
| MFCD03791313 | |
| Soluble in DMSO or 100% ethanol. | |
| CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCC(=O)O | |
| 5 mg | |
| 5283389 | |
| 361.52 | |
| Waxy Solid |
| 179113-91-8 | |
| C22H35NO3 | |
| n-arachidonylglycine, n-arachidonoylglycine, nagly, n-arachidonoyl glycine, arachidonoyl glycine, n-5z,8z,11z,14z-eicosatetraenoyl-glycine, dsstox_cid_25124, dsstox_rid_80688, dsstox_gsid_45124, 2-5z,8z,11z,14z-icosa-5,8,11,14-tetraenoyl amino acetic acid | |
| YLEARPUNMCCKMP-DOFZRALJSA-N | |
| 2-[[(5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoyl]amino]acetic acid | |
| 361.526 | |
| CHEBI:58961 | |
| Air sensitive |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen