accessibility menu, dialog, popup

UserName

CAS RN 179113-91-8

CH3ONHOOH(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 179113-91-8

IUPAC Name: 2-(icosa-5,8,11,14-tetraenamido)acetic acid
Synonyms: N-arachidonylglycine
Molecular Weight (g/mol): 361.53
Molecular Formula: C22H35NO3
InChi Key: YLEARPUNMCCKMP-UHFFFAOYSA-N
SMILES: CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)