missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N(alpha)-Boc-N(epsilon),N(epsilon)-dimethyl-L-lysine, 97%
CAS: 65671-53-6 | C13H26N2O4 | 274.36 g/mol
Supplier: Thermo Scientific Chemicals H5243703
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 65671-53-6 | |
| 274.36 | |
| KPXRFYHPDPDJSY-UEQNJFAPNA-N | |
| 7018778 | |
| CN(C)CCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O |
| C13H26N2O4 | |
| MFCD00076965 | |
| boc-lys me 2-oh, l-lysine,n2-1,1-dimethylethoxy carbonyl-n6,n6-dimethyl, 2s-2-tert-butoxycarbonyl amino-6-dimethylamino hexanoic acid, boc-lys me-oh, nalpha-boc-nepsilon-dimethyl-l-lysine, n-alpha-boc-s-2-amino-6-dimethylamino hexanoic acid, 2 s-+-tert-butoxycarbonylamino-6-dimethylaminohexanoic acid, s-2-tert-butoxycarbonylamino-6-dimethylamino hexanoic acid, 2s-2-tert-butoxy carbonyl amino-6-dimethylamino hexanoic acid | |
| (2S)-6-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Specifications
| N(alpha)-Boc-N(epsilon),N(epsilon)-dimethyl-L-lysine | |
| C13H26N2O4 | |
| MFCD00076965 | |
| KPXRFYHPDPDJSY-UEQNJFAPNA-N | |
| (2S)-6-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 7018778 | |
| 97% |
| 65671-53-6 | |
| 1 g | |
| boc-lys me 2-oh, l-lysine,n2-1,1-dimethylethoxy carbonyl-n6,n6-dimethyl, 2s-2-tert-butoxycarbonyl amino-6-dimethylamino hexanoic acid, boc-lys me-oh, nalpha-boc-nepsilon-dimethyl-l-lysine, n-alpha-boc-s-2-amino-6-dimethylamino hexanoic acid, 2 s-+-tert-butoxycarbonylamino-6-dimethylaminohexanoic acid, s-2-tert-butoxycarbonylamino-6-dimethylamino hexanoic acid, 2s-2-tert-butoxy carbonyl amino-6-dimethylamino hexanoic acid | |
| CN(C)CCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
| 274.36 | |
| 274.36 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures