missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Boc-N(epsilon),N(epsilon)-dimethyl-L-lysine, 97%

Catalog No. p-7034739
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52437MD 250 mg
AAH5243703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52437MD Supplier Thermo Scientific Chemicals Supplier No. H52437MD

CAS: 65671-53-6 | C13H26N2O4 | 274.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65671-53-6
Molecular Formula C13H26N2O4
Molecular Weight (g/mol) 274.36
MDL Number MFCD00076965
InChI Key KPXRFYHPDPDJSY-UEQNJFAPNA-N
Synonym boc-lys me 2-oh, l-lysine,n2-1,1-dimethylethoxy carbonyl-n6,n6-dimethyl, 2s-2-tert-butoxycarbonyl amino-6-dimethylamino hexanoic acid, boc-lys me-oh, nalpha-boc-nepsilon-dimethyl-l-lysine, n-alpha-boc-s-2-amino-6-dimethylamino hexanoic acid, 2 s-+-tert-butoxycarbonylamino-6-dimethylaminohexanoic acid, s-2-tert-butoxycarbonylamino-6-dimethylamino hexanoic acid, 2s-2-tert-butoxy carbonyl amino-6-dimethylamino hexanoic acid
PubChem CID 7018778
IUPAC Name (2S)-6-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CN(C)CCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 274.36
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-N(epsilon),N(epsilon)-dimethyl-L-lysine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only