Learn More
N(alpha)-Boc-D-2,3-diaminopropionic acid, 97%
CAS: 76387-70-7 | C8H16N2O4 | 204.226 g/mol
Supplier: Thermo Scientific Chemicals H5282103
Description
N(alpha)-Boc-D-2,3-diaminopropionic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN(alpha)-Boc-D-2,3-diaminopropionic acid is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
| N(alpha)-Boc-D-2,3-diaminopropionic acid | |
| 200°C to 203°C | |
| MFCD01632072 | |
| boc-d-dap-oh, boc-d-2,3-diaminopropionic acid, r-3-amino-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-amino-2-tert-butoxy carbonyl amino propanoic acid, d-alanine, 3-amino-n-1,1-dimethylethoxy carbonyl, 2r-3-amino-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-amino-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, ambotzbaa1179, pubchem14749, 3-amino-n-boc-d-alanine | |
| +14° (c=1 in Water) | |
| (2R)-3-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 7020984 | |
| 97% |
| 76387-70-7 | |
| C8H16N2O4 | |
| 1 g | |
| KRJLRVZLNABMAT-RXMQYKEDSA-N | |
| CC(C)(C)OC(=O)NC(CN)C(=O)O | |
| 204.226 | |
| 204.23 |
Chemical Identifiers
| 76387-70-7 | |
| 204.226 | |
| KRJLRVZLNABMAT-RXMQYKEDSA-N | |
| 7020984 | |
| CC(C)(C)OC(=O)NC(CN)C(=O)O |
| C8H16N2O4 | |
| MFCD01632072 | |
| boc-d-dap-oh, boc-d-2,3-diaminopropionic acid, r-3-amino-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-amino-2-tert-butoxy carbonyl amino propanoic acid, d-alanine, 3-amino-n-1,1-dimethylethoxy carbonyl, 2r-3-amino-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-amino-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, ambotzbaa1179, pubchem14749, 3-amino-n-boc-d-alanine | |
| (2R)-3-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only