missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-D-2,3-diaminopropionic acid, 97%

Catalog No. p-7035113
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52821MD 250 mg
AAH5282103 1 g
AAH5282106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52821MD Supplier Thermo Scientific Chemicals Supplier No. H52821MD
Only null left

CAS: 76387-70-7 | C8H16N2O4 | 204.226 g/mol

N(alpha)-Boc-D-2,3-diaminopropionic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N(alpha)-Boc-D-2,3-diaminopropionic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 76387-70-7
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.226
MDL Number MFCD01632072
InChI Key KRJLRVZLNABMAT-RXMQYKEDSA-N
Synonym boc-d-dap-oh, boc-d-2,3-diaminopropionic acid, r-3-amino-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-amino-2-tert-butoxy carbonyl amino propanoic acid, d-alanine, 3-amino-n-1,1-dimethylethoxy carbonyl, 2r-3-amino-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-amino-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, ambotzbaa1179, pubchem14749, 3-amino-n-boc-d-alanine
PubChem CID 7020984
IUPAC Name (2R)-3-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CN)C(=O)O

Specifications

Melting Point 200°C to 203°C
Quantity 250 mg
Optical Rotation +14° (c=1 in Water)
Formula Weight 204.23
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-D-2,3-diaminopropionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only