missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-Benzoyl-L-arginine, 99%

Catalog No. AAA1081209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1081209 10 g
AAA1081218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1081209 Supplier Thermo Scientific Chemicals Supplier No. A1081209

CAS: 154-92-7 | C13H18N4O3 | 278.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154-92-7
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.31
MDL Number MFCD00001763
InChI Key RSYYQCDERUOEFI-UHFFFAOYNA-N
Synonym bz-arg-oh, n-alpha-benzoyl-l-arginine, n-benzoyl-l-arginine, n2-benzoyl-l-arginine, benzoyl-l-arginine, arginine, n2-benzoyl, nalpha-benzoyl-l-arginine, 2s-5-carbamimidamido-2-phenylformamido pentanoic acid, l-arginine, n2-benzoyl, n∼2∼-phenylcarbonyl-l-arginine
PubChem CID 97369
IUPAC Name (2S)-2-benzamido-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 285°C (decomposition)
Quantity 10 g
Formula Weight 278.31
Percent Purity 99%
Chemical Name or Material N-alpha-Benzoyl-L-arginine
EINECSNumber 205-837-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only