accessibility menu, dialog, popup

UserName

CAS RN 154-92-7

IUPAC Name: 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanoic acid
Synonyms: 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanoic acid
Molecular Weight (g/mol): 278.31
Molecular Formula: C13H18N4O3
InChi Key: RSYYQCDERUOEFI-UHFFFAOYNA-N
SMILES: NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)